missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-17B Recombinant Protein, (CHO-expressed)

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 27190
Molecular Weight 18 kDa (monomer)
Name IL-17B
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
8129IL025CF R&D Systems
RDS8129IL025CF
Lyophilized from a 0.2μm filtered solution in Sodium Phosphate and NaCl. Each for $416.79
8129IL025 R&D Systems
RDS8129IL025
Lyophilized from a 0.2μm filtered solution in Sodium Phosphate and NaCl with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Gln21-Phe180
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Sodium Phosphate and NaCl.
Entrez Gene ID: 27190
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
27190
IL-17B
25μg
Human