Please login to your online account to display your discounted pricing

R&D Systems Human IL-17B Recombinant Protein

$407.05 - $407.05

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 27190
Molecular Weight 18.3 kDa
Name IL-17B
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1248IB025

 
r&d systems
RDS1248IB025
Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl with BSA as a carrier protein. Each for $407.05

1248IB025CF

 
r&d systems
RDS1248IB025CF
Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl. Each for $407.05
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 27190
Molecular Weight 18.3 kDa
Name IL-17B
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: E. coli-derived, Gln21-Phe180, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl.
Entrez Gene ID: 27190
Regulatory Status: RUO