missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-17A/F Heterodimer Recombinant Protein

$456.25 - $456.25

Specifications

Conjugate Unlabeled
Molecular Weight IL-17A: 15.7 kDa and IL-17F: 15 kDa
Name IL-17A/F Heterodimer
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
5194IL025 R&D Systems
RDS5194IL025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $456.25
5194IL025CF R&D Systems
RDS5194IL025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $456.25
Specifications

Description

Source: E. coli-derived, Ile20-Ala155 (IL-17A) & Arg31-Gln163 (IL-17F)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: IL-17A: 15.7 kDa and IL-17F: 15 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IL-17A/F Heterodimer
25μg
Human
IL-17A: 15.7 kDa and IL-17F: 15 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes