missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-17/IL-17A Recombinant Protein, (Human Cell-expressed)

$337.62 - $337.62

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3605
Molecular Weight 15.1 kDa (monomer)
Name IL-17/IL-17A
Purity 95%, by SDS-PAGE with silver staining.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
7955IL025 R&D Systems
RDS7955IL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $337.62
7955IL025CF R&D Systems
RDS7955IL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $337.62
Specifications

Description

Source: Human embryonic kidney cell, HEK293-derived, Gly24-Ala155
Purity: 95%, by SDS-PAGE with silver staining.
Molecular Weight: 15.1 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3605
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15.1 kDa (monomer)
95%, by SDS-PAGE with silver staining.
Yes
3605
IL-17/IL-17A
25μg
Human