missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-15 Recombinant Protein

$290.11 - $842.45

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3600
Molecular Weight 12.5 kDa
Name IL-15
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
247IL005 R&D Systems
RDS247IL005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $290.11
247IL025 R&D Systems
RDS247IL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $842.45
Specifications

Description

Source: E. coli-derived, Asn49-Ser162
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3600
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
12.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
3600
IL-15
Yes