missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-13 Recombinant Protein

$389.76 - $7,430.30

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3596
Molecular Weight 12.3 kDa
Name IL-13
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
213ILB005 R&D Systems
RDS213ILB005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $389.76
213ILB01MGC R&D Systems
RDS213ILB01MGCF
Lyophilized from a 0.2μm filtered solution in PBS. 1mg Each for $7,430.30
213ILB025 R&D Systems
RDS213ILB025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $1,231.87
213ILB005CF R&D Systems
RDS213ILB005CF
Lyophilized from a 0.2μm filtered solution in PBS. 5μg Each for $389.76
213ILB025CF R&D Systems
RDS213ILB025CF
Lyophilized from a 0.2μm filtered solution in PBS. 25μg Each for $1,231.87
Specifications

Description

Source: E. coli-derived, Gly21-Asn132
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3596
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
12.3 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
3596
IL-13
Yes