Please login to your online account to display your discounted pricing

R&D Systems Human IL-10 Recombinant Protein, (aa 19-178)


Manufacturer: r&d systems  RDS1064IL010CF

 View more versions of this product

Catalog No. 1064IL010CF

  • / Each
 

Description & Specifications

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3586
Molecular Weight 18.8 kDa
Name IL-10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Source Human
Structural Form Noncovalently-linked homodimer
Conjugate Unlabeled
Recombinant Yes