missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-10 Recombinant Protein, (aa 19-178)

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3586
Molecular Weight 18.8 kDa
Name IL-10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1064IL010 R&D Systems
RDS1064IL010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $376.73
1064IL010CF R&D Systems
RDS1064IL010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $376.73
Specifications

Description

Source: E. coli-derived, Ser19-Asn178, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3586
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3586
IL-10
10μg
Human