missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-10 Recombinant Protein, Ser19-Asn178

$389.76 - $1,166.69

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3586
Molecular Weight 18.6 kDa
Name IL-10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
217IL005CF R&D Systems
RDS217IL005CF
Lyophilized from a 0.2μm filtered solution in PBS. 5μg Each for $389.76
217IL005 R&D Systems
RDS217IL005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $389.76
217IL025 R&D Systems
RDS217IL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $1,166.69
217IL025CF R&D Systems
RDS217IL025CF
Lyophilized from a 0.2μm filtered solution in PBS. 25μg Each for $1,166.69
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ser19-Asn178
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3586
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
3586
IL-10
Yes