missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-1 RII Recombinant Protein

$533.15 - $533.15

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7850
Molecular Weight 36 kDa
Name IL-1 RII
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2632R050 R&D Systems
RDS2632R050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $533.15
2632R050CF R&D Systems
RDS2632R050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $533.15
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Phe14-Glu343
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 7850
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
36 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
7850
IL-1 RII
50μg
Human