missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-1 RI Recombinant Protein

$663.51 - $663.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3554
Molecular Weight 36 kDa
Name IL-1 RI
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2691R100 R&D Systems
RDS2691R100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $663.51
2691R100CF R&D Systems
RDS2691R100CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $663.51
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Asp21-Thr332
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3554
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
36 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3554
IL-1 RI
100μg
Human