missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-1 alpha/IL-1F1 Recombinant Protein

$233.33 - $793.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3552
Molecular Weight 18 kDa
Name IL-1 alpha/IL-1F1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
200LA002 R&D Systems
RDS200LA002
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 2μg Each for $233.33
200LA002CF R&D Systems
RDS200LA002CF
Supplied as a 0.2μm filtered solution in PBS. 2μg Each for $233.33
200LA010 R&D Systems
RDS200LA010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10μg Each for $793.87
200LA010CF R&D Systems
RDS200LA010CF
Supplied as a 0.2μm filtered solution in PBS. 10μg Each for $793.87
Specifications

Description

Source: E. coli-derived, Ser113-Ala271
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Supplied as a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3552
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
3552
IL-1 alpha/IL-1F1
Yes