missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IGF-II Recombinant Protein

$233.33 - $814.73

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in Acetic Acid.
Gene ID (Entrez) 3481
Molecular Weight 7.5 kDa
Name IGF-II
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
292G2050 R&D Systems
RDS292G2050
50μg Each for $233.33
292G2250 R&D Systems
RDS292G2250
250μg Each for $814.73
Specifications

Description

Source: E. coli-derived, Ala25-Glu91
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetic Acid.
Entrez Gene ID: 3481
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
3481
IGF-II
Yes
Lyophilized from a 0.2μm filtered solution in Acetic Acid.
7.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human