Please login to your online account to display your discounted pricing

R&D Systems Human IGF-I Recombinant Protein

$259.40 - $767.80

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3479
Molecular Weight 7.6 kDa
Name IGF-I
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

291G101M

 
r&d systems
RDS291G101M
1mg Each for $767.80

291G1200

 
r&d systems
RDS291G1200
200μg Each for $259.40
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3479
Molecular Weight 7.6 kDa
Name IGF-I
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Gly49-Ala118
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3479
Regulatory Status: RUO