Please login to your online account to display your discounted pricing

R&D Systems Human IFN-alpha A/D [BglII] (Universal Type I Interferon) Recombinant Protein

$444.56 - $768.68

Specifications

Formulation Phosphate buffered saline (PBS) containing 0.1% bovine serum albumin (BSA)
Purification Method Purified from E. coli by a combination of hydrophobic interaction, ion exchange and gel permeation chromatography
Purity >95%
Recombinant Yes
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

112002

 
r&d systems
112002
1MU Each for $768.68

112001

 
r&d systems
112001
100000U Each for $444.56
Description & Specifications

Specifications

Formulation Phosphate buffered saline (PBS) containing 0.1% bovine serum albumin (BSA)
Purification Method Purified from E. coli by a combination of hydrophobic interaction, ion exchange and gel permeation chromatography
Purity >95%
Recombinant Yes