missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human ICAM-3/CD50 Fc Chimera Recombinant Protein

$290.11 - $860.18

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3385
Molecular Weight 76.7 kDa (monomer)
Name ICAM-3/CD50 Fc Chimera
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
715IC200 R&D Systems
RDS715IC200
200μg Each for $860.18
715IC050 R&D Systems
RDS715IC050
50μg Each for $290.11
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gln30-His485
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 76.7 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3385
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
3385
ICAM-3/CD50 Fc Chimera
Yes
Lyophilized from a 0.2μm filtered solution in PBS.
76.7 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human