Please login to your online account to display your discounted pricing

R&D Systems Human ICAM-2/CD102 Fc Chimera Recombinant Protein

$298.51 - $885.12

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol.
Gene ID (Entrez) 3384
Molecular Weight 50 kDa (monomer)
Name ICAM-2/CD102 Fc Chimera
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

803I2050

 
r&d systems
RDS803I2050
50μg Each for $298.51

803I2200

 
r&d systems
RDS803I2200
200μg Each for $885.12
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol.
Gene ID (Entrez) 3384
Molecular Weight 50 kDa (monomer)
Name ICAM-2/CD102 Fc Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, Lys25-Gln223
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 50 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol.
Entrez Gene ID: 3384
Regulatory Status: RUO