missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human ICAM-1/CD54 Recombinant Protein

$423.65 - $997.23

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Trehalose.
Gene ID (Entrez) 3383
Molecular Weight 50 kDa
Name ICAM-1/CD54
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ADP4050 R&D Systems
RDSADP4050
50μg Each for $423.65
ADP4200 R&D Systems
RDSADP4200
200μg Each for $997.23
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Met1-Glu480
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 50 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Trehalose.
Entrez Gene ID: 3383
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
3383
ICAM-1/CD54
Yes
Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Trehalose.
50 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human