Please login to your online account to display your discounted pricing

R&D Systems Human ICAM-1/CD54 Fc Chimera Recombinant Protein

$292.21 - $866.42

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3383
Molecular Weight 76 kDa (monomer)
Name ICAM-1/CD54 Fc Chimera
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

720IC050

 
r&d systems
RDS720IC050
50μg Each for $292.21

720IC200

 
r&d systems
RDS720IC200
200μg Each for $866.42
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3383
Molecular Weight 76 kDa (monomer)
Name ICAM-1/CD54 Fc Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, Gln28-Glu480
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 76 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3383
Regulatory Status: RUO