missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human HVEM/TNFRSF14 Fc Chimera Recombinant Protein

$454.94 - $454.94

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 8764
Molecular Weight 45 kDa (monomer)
Name HVEM/TNFRSF14 Fc Chimera
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
356HV100 R&D Systems
RDS356HV100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $454.94
356HV100CF R&D Systems
RDS356HV100CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $454.94
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Pro37-Val202 (Ser108Thr and Ala140Arg)
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 45 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 8764
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
45 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
8764
HVEM/TNFRSF14 Fc Chimera
100μg
Human