Please login to your online account to display your discounted pricing

R&D Systems Human HSP70/HSPA1A Recombinant Protein

$369.17 - $460.42

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 3303
Molecular Weight 70 kDa
Name HSP70/HSPA1A
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AP100050

 
r&d systems
RDSAP100050
50μg Each for $369.17

AP100100

 
r&d systems
RDSAP100100
100μg Each for $460.42
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 3303
Molecular Weight 70 kDa
Name HSP70/HSPA1A
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight: 70 kDa
Formulation: Supplied as a solution in Tris, NaCl and Glycerol.
Entrez Gene ID: 3303
Regulatory Status: RUO