Please login to your online account to display your discounted pricing

R&D Systems Human His6 Use1/UBE2Z Recombinant Protein


Manufacturer: r&d systems  RDSE2677050

 View more versions of this product

Catalog No. E2677050

  • / Each
 

Description & Specifications

Specifications

Name His6 Use1/UBE2Z
Source Human
Formulation No Formulation Info in Bulk Inventory
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Gene ID (Entrez) 65264
Recombinant Yes
Quantity 50μg
Conjugate Unlabeled