Please login to your online account to display your discounted pricing

R&D Systems Human His6 Ufc1 Recombinant Protein

$172.26 - $286.85

Specifications

Conjugate Unlabeled
Formulation No Formulation Info in Bulk Inventory
Gene ID (Entrez) 51506
Name His6 Ufc1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2675050

 
r&d systems
RDSE2675050
50μg Each for $172.26

E2675100

 
r&d systems
RDSE2675100
100μg Each for $286.85
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation No Formulation Info in Bulk Inventory
Gene ID (Entrez) 51506
Name His6 Ufc1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: No Formulation Info in Bulk Inventory
Entrez Gene ID: 51506
Regulatory Status: RUO