Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2W Isoform 2 Recombinant Protein


Manufacturer: r&d systems  RDSE2725100

 View more versions of this product

Catalog No. E2725100

  • / Each
 

Description & Specifications

Specifications

Accession Number Q96B02-2
Name His6 UBE2W Isoform 2
Source Human
Gene ID (Entrez) 55284
Recombinant Yes
Conjugate Unlabeled
Quantity 100μg