Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2W Isoform 2 Recombinant Protein


Manufacturer: r&d systems  RDSE2725050

 View more versions of this product

Catalog No. E2725050

  • / Each
 

Description & Specifications

Specifications

Quantity 50μg
Name His6 UBE2W Isoform 2
Accession Number Q96B02-2
Source Human
Recombinant Yes
Conjugate Unlabeled
Gene ID (Entrez) 55284