Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2W Isoform 2 Recombinant Protein

$172.26 - $286.85

Specifications

Accession Number Q96B02-2
Conjugate Unlabeled
Gene ID (Entrez) 55284
Name His6 UBE2W Isoform 2
Recombinant Yes
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2725050

 
r&d systems
RDSE2725050
50μg Each for $172.26

E2725100

 
r&d systems
RDSE2725100
100μg Each for $286.85
Description & Specifications

Specifications

Accession Number Q96B02-2
Conjugate Unlabeled
Gene ID (Entrez) 55284
Name His6 UBE2W Isoform 2
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal 6-His tag
Entrez Gene ID: 55284
Regulatory Status: RUO