Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2W Isoform 1 Recombinant Protein

Manufacturer: r&d systems  RDSE2740050

 View all options for this product

Catalog No. E2740050


Description & Specifications


Conjugate Unlabeled
Gene ID (Entrez) 55284
Name His6 UBE2W Isoform 1
Quantity 50μg
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal 6-His tag
Entrez Gene ID: 55284
Regulatory Status: RUO