missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2W Isoform 1 Recombinant Protein

$293.04 - $293.04

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 55284
Name His6 UBE2W Isoform 1
Recombinant Yes
Source Human
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E2740100 R&D Systems
RDSE2740100
100μg Each for $293.04
Specifications

Description

Source: E. coli-derived, Contains an N-terminal 6-His tag
Entrez Gene ID: 55284
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
His6 UBE2W Isoform 1
Human
55284
Yes