Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2W Isoform 1 Recombinant Protein


Manufacturer: r&d systems  RDSE2740100

 View more versions of this product

Catalog No. E2740100

  • / Each
 

Description & Specifications

Specifications

Source Human
Gene ID (Entrez) 55284
Name His6 UBE2W Isoform 1
Quantity 100μg
Conjugate Unlabeled
Recombinant Yes