Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2S Recombinant Protein


Manufacturer: r&d systems  RDSE2690050

 View more versions of this product

Catalog No. E2690050

  • / Each
 

Description & Specifications

Specifications

Name His6 UBE2S
Formulation No Formulation Info in Bulk Inventory
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Quantity 50μg
Recombinant Yes
Source Human
Conjugate Unlabeled
Gene ID (Entrez) 27338