Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2R2 Recombinant Protein


Manufacturer: r&d systems  RDSE2715050

 View more versions of this product

Catalog No. E2715050

  • / Each
 

Description & Specifications

Specifications

Source Human
Gene ID (Entrez) 54926
Accession Number Q712K3
Quantity 50μg
Recombinant Yes
Conjugate Unlabeled
Name His6 UBE2R2