Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2R2 Recombinant Protein

$172.26 - $286.85

Specifications

Accession Number Q712K3
Conjugate Unlabeled
Gene ID (Entrez) 54926
Name His6 UBE2R2
Recombinant Yes
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2715050

 
r&d systems
RDSE2715050
50μg Each for $172.26

E2715100

 
r&d systems
RDSE2715100
100μg Each for $286.85
Description & Specifications

Specifications

Accession Number Q712K3
Conjugate Unlabeled
Gene ID (Entrez) 54926
Name His6 UBE2R2
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal 6-His tag
Entrez Gene ID: 54926
Regulatory Status: RUO