Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2R2 Recombinant Protein


Manufacturer: r&d systems  RDSE2715100

 View more versions of this product

Catalog No. E2715100

  • / Each
 

Description & Specifications

Specifications

Accession Number Q712K3
Name His6 UBE2R2
Quantity 100μg
Recombinant Yes
Source Human
Conjugate Unlabeled
Gene ID (Entrez) 54926