Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2Q2 Isoform 1 Recombinant Protein


Manufacturer: r&d systems  RDSE2745050

 View more versions of this product

Catalog No. E2745050

  • / Each
 

Description & Specifications

Specifications

Source Human
Gene ID (Entrez) 92912
Name His6 UBE2Q2 Isoform 1
Accession Number Q8WVN8
Recombinant Yes
Conjugate Unlabeled
Quantity 50μg