Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2Q2 Isoform 1 Recombinant Protein

$175.98 - $293.04

Specifications

Accession Number Q8WVN8
Conjugate Unlabeled
Gene ID (Entrez) 92912
Name His6 UBE2Q2 Isoform 1
Recombinant Yes
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2745050

 
r&d systems
RDSE2745050
50μg Each for $175.98

E2745100

 
r&d systems
RDSE2745100
100μg Each for $293.04
Description & Specifications

Specifications

Accession Number Q8WVN8
Conjugate Unlabeled
Gene ID (Entrez) 92912
Name His6 UBE2Q2 Isoform 1
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal 6-His tag
Entrez Gene ID: 92912
Regulatory Status: RUO