Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2Q1 Isoform 2 Recombinant Protein


Manufacturer: r&d systems  RDSE2720050

 View more versions of this product

Catalog No. E2720050

  • / Each
 

Description & Specifications

Specifications

Name His6 UBE2Q1 Isoform 2
Quantity 50μg
Source Human
Conjugate Unlabeled
Accession Number Q7Z7E8-2
Gene ID (Entrez) 55585
Recombinant Yes