Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2Q1 Isoform 2 Recombinant Protein

$175.98 - $293.04

Specifications

Accession Number Q7Z7E8-2
Conjugate Unlabeled
Gene ID (Entrez) 55585
Name His6 UBE2Q1 Isoform 2
Recombinant Yes
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2720100

 
r&d systems
RDSE2720100
100μg Each for $293.04

E2720050

 
r&d systems
RDSE2720050
50μg Each for $175.98
Description & Specifications

Specifications

Accession Number Q7Z7E8-2
Conjugate Unlabeled
Gene ID (Entrez) 55585
Name His6 UBE2Q1 Isoform 2
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal 6-His tag
Entrez Gene ID: 55585
Regulatory Status: RUO