Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2Q1 Isoform 1 Recombinant Protein


Manufacturer: r&d systems  RDSE2755050

 View more versions of this product

Catalog No. E2755050

  • / Each
 

Description & Specifications

Specifications

Accession Number Q7Z7E8
Quantity 50μg
Recombinant Yes
Source Human
Conjugate Unlabeled
Gene ID (Entrez) 55585
Name His6 UBE2Q1 Isoform 1