Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2Q1 Isoform 1 Recombinant Protein


Manufacturer: r&d systems  RDSE2755100

 View more versions of this product

Catalog No. E2755100

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 55585
Conjugate Unlabeled
Accession Number Q7Z7E8
Name His6 UBE2Q1 Isoform 1
Recombinant Yes
Quantity 100μg
Source Human