Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2J2 Isoform 1 Recombinant Protein


Manufacturer: r&d systems  RDSE2710100

 View more versions of this product

Catalog No. E2710100

  • / Each
 

Description & Specifications

Specifications

Name His6 UBE2J2 Isoform 1
Gene ID (Entrez) 118424
Accession Number Q8N2K1
Quantity 100μg
Conjugate Unlabeled
Source Human
Recombinant Yes