Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2J2 Isoform 1 Recombinant Protein

$172.26 - $286.85

Specifications

Accession Number Q8N2K1
Conjugate Unlabeled
Gene ID (Entrez) 118424
Name His6 UBE2J2 Isoform 1
Recombinant Yes
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2710050

 
r&d systems
RDSE2710050
50μg Each for $172.26

E2710100

 
r&d systems
RDSE2710100
100μg Each for $286.85
Description & Specifications

Specifications

Accession Number Q8N2K1
Conjugate Unlabeled
Gene ID (Entrez) 118424
Name His6 UBE2J2 Isoform 1
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal 6-His tag
Entrez Gene ID: 118424
Regulatory Status: RUO