Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2J1 Recombinant Protein


Manufacturer: r&d systems  RDSE2750050

 View more versions of this product

Catalog No. E2750050

  • / Each
 

Description & Specifications

Specifications

Accession Number Q9Y385
Name His6 UBE2J1
Quantity 50μg
Recombinant Yes
Source Human
Conjugate Unlabeled
Gene ID (Entrez) 51465