Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2J1 Recombinant Protein

$175.98 - $293.04

Specifications

Accession Number Q9Y385
Conjugate Unlabeled
Gene ID (Entrez) 51465
Name His6 UBE2J1
Recombinant Yes
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2750050

 
r&d systems
RDSE2750050
50μg Each for $175.98

E2750100

 
r&d systems
RDSE2750100
100μg Each for $293.04
Description & Specifications

Specifications

Accession Number Q9Y385
Conjugate Unlabeled
Gene ID (Entrez) 51465
Name His6 UBE2J1
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal 6-His tag and the C-terminal transmembrane and lumenal amino acid sequences have been removed
Entrez Gene ID: 51465
Regulatory Status: RUO