Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2G2 Recombinant Protein


Manufacturer: r&d systems  RDSE2680100

 View more versions of this product

Catalog No. E2680100

  • / Each
 

Description & Specifications

Specifications

Name His6 UBE2G2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Quantity 100μg
Recombinant Yes
Source Human
Conjugate Unlabeled
Formulation Formulation cannot be determined without the BulkLotPrefix
Gene ID (Entrez) 7327