Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2G1 Recombinant Protein


Manufacturer: r&d systems  RDSE2700050

 View more versions of this product

Catalog No. E2700050

  • / Each
 

Description & Specifications

Specifications

Name His6 UBE2G1
Quantity 50μg
Recombinant Yes
Source Human
Conjugate Unlabeled
Gene ID (Entrez) 7326