Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2F Recombinant Protein

$175.98 - $293.04

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 140739
Name His6 UBE2F
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2679100

 
r&d systems
RDSE2679100
100μg Each for $293.04

E2679050

 
r&d systems
RDSE2679050
50μg Each for $175.98
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 140739
Name His6 UBE2F
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 140739
Regulatory Status: RUO