missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2E3 Recombinant Protein

$175.98 - $293.04

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 10477
Name His6 UBE2E3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E2678100 R&D Systems
RDSE2678100
100μg Each for $293.04
E2678050 R&D Systems
RDSE2678050
50μg Each for $175.98
Specifications

Description

Source: E. coli-derived, Contains an N-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 10477
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
10477
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Human
Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
His6 UBE2E3
Yes