Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2E3 Recombinant Protein


Manufacturer: r&d systems  RDSE2678100

 View more versions of this product

Catalog No. E2678100

  • / Each
 

Description & Specifications

Specifications

Name His6 UBE2E3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Source Human
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 10477
Quantity 100μg
Recombinant Yes
Conjugate Unlabeled