Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2D4/Ubch5d Recombinant Protein


Manufacturer: r&d systems  RDSE2705100

 View more versions of this product

Catalog No. E2705100

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 51619
Name His6 UBE2D4/Ubch5d
Accession Number Q9Y2X8
Source Human
Recombinant Yes
Conjugate Unlabeled
Quantity 100μg