Please login to your online account to display your discounted pricing

R&D Systems Human His6 UBE2D4/Ubch5d Recombinant Protein

$172.26 - $286.85

Specifications

Accession Number Q9Y2X8
Conjugate Unlabeled
Gene ID (Entrez) 51619
Name His6 UBE2D4/Ubch5d
Recombinant Yes
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2705100

 
r&d systems
RDSE2705100
100μg Each for $286.85

E2705050

 
r&d systems
RDSE2705050
50μg Each for $172.26
Description & Specifications

Specifications

Accession Number Q9Y2X8
Conjugate Unlabeled
Gene ID (Entrez) 51619
Name His6 UBE2D4/Ubch5d
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal 6-His tag
Entrez Gene ID: 51619
Regulatory Status: RUO